Skip to Content
Merck
All Photos(1)

Key Documents

140287

Sigma-Aldrich

1,2-Diphenoxyethane

1,2-Diphenoxyethane

99%

Synonym(s):

Ethylene glycol diphenyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€54.40

€54.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€54.40

About This Item

Linear Formula:
C6H5OCH2CH2OC6H5
CAS Number:
Molecular Weight:
214.26
Beilstein:
2052248
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€54.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

94-96 °C (lit.)

functional group

phenoxy

SMILES string

C(COc1ccccc1)Oc2ccccc2

InChI

1S/C14H14O2/c1-3-7-13(8-4-1)15-11-12-16-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

XCSGHNKDXGYELG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Diphenoxyethane is the intermediate formed during sonochemical oxidation of phenoxyacetic acid and chlorophenol[1].

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

341.2 °F

Flash Point(C)

171.8 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yakov Yasman et al.
Ultrasonics sonochemistry, 13(3), 271-277 (2005-06-25)
The application of finely divided (black) Pd and Pd-Fe powder in the sono-electro-catalytic reduction of chlorophenoxy herbicides (2,4-D) and chlorophenols (2,4-DCP) in aqueous solutions allows for effective destruction of toxic chlorinated aromatic compounds. At 20 degrees C complete conversion of
Martin Eckardt et al.
Journal of chromatography. A, 1609, 460437-460437 (2019-08-15)
Thermal papers (e.g. point of sale receipts, adhesive labels, tickets) significantly contribute to contamination of paper material cycles and the environment with substances of (eco-) toxicological concern. In particular, they contain color developers like endocrine disrupting bisphenols in typical concentrations

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service