Skip to Content
Merck
All Photos(1)

Documents

139130

Sigma-Aldrich

Methyl 2-chloropropionate

97%

Synonym(s):

(±)-Methyl 2-chloropropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHClCOOCH3
CAS Number:
Molecular Weight:
122.55
Beilstein:
1421535
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.22 (vs air)

vapor pressure

5.25 mmHg ( 20 °C)

Assay

97%

expl. lim.

19 %

refractive index

n20/D 1.417 (lit.)

bp

132-133 °C (lit.)

density

1.075 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(C)Cl

InChI

1S/C4H7ClO2/c1-3(5)4(6)7-2/h3H,1-2H3

InChI key

JLEJCNOTNLZCHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 2-chloropropionate is the initiating/catalyst system during the atom transfer radical polymerization of N,N-dimethylacrylamide, N-isopropylacrylamide and N-vinylpyrrolidone.

Application

Methyl 2-chloropropionate was used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A I Gurova et al.
Gigiena truda i professional'nye zabolevaniia, (5)(5), 36-38 (1992-01-01)
Reports of MACs for carboxylic acid derivatives as sodium methoxyacetate (SMOA), methyl chlorpropionate (MCP) and acetoacetic ether (AAE). SMOA shows low toxicity, no cumulation and specific action MCP demonstrates moderate toxicity, low cumulation no specific action in concentrations close to
Controllable synthesis of poly (N-vinylpyrrolidone) and its block copolymers by atom transfer radical polymerization.
Lu X, et al.
Polymer, 48(10), 2835-2842 (2007)
Thermal response of narrow-disperse poly (N-isopropylacrylamide) prepared by atom transfer radical polymerization.
Xia Y, et al.
Macromolecules, 38(14), 5937-5943 (2005)
Acrylamide Homopolymers and Acrylamide-N-Isopropylacrylamide Block Copolymers by Atomic Transfer Radical Polymerization in Water.
Wever DAZ, et al.
Macromolecules, 45(10), 4040-4045 (2012)
Copolymerization of N, N-dimethylacrylamide with n-butyl acrylate via atom transfer radical polymerization.
Neugebauer D and Matyjaszewski K.
Macromolecules, 36(8), 2598-2603 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service