Skip to Content
Merck
All Photos(1)

Key Documents

137863

Sigma-Aldrich

3,5-Dimethylaniline

98%

Synonym(s):

1-Amino-3,5-dimethylbenzene, 3,5-Xylidine, 5-Amino-m-xylene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€62.00
500 ML
€206.00

€62.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€62.00
500 ML
€206.00

About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507051
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€62.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.557 (lit.)

bp

104-105 °C/14 mmHg (lit.)

mp

7-9 °C

density

0.972 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)cc(N)c1

InChI

1S/C8H11N/c1-6-3-7(2)5-8(9)4-6/h3-5H,9H2,1-2H3

InChI key

MKARNSWMMBGSHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5-Dimethylaniline is the reagent commonly useful in the manufacture of dyes[1]. It undergoes polymerization in the presence of cerium(IV) sulfate as an oxidant[2].

Application

3,5-Dimethylaniline was used in the synthesis of chiral packing materials for high performance liquid chromatography[3].

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

215.1 °F - closed cup

Flash Point(C)

101.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemical preparation of polyaniline and its derivatives by using cerium (IV) sulfate.
Yan H and Toshima N.
Synt. Metals, 69(1), 151-152 (1995)
The enantiomeric recognition of dihydropyrimidonic compounds by chiral selectors derived from 4-or 2-chloro-3, 5-dinitrobenzoic acid.
Gazic I, et al.
Tetrahedron Asymmetry, 16(6), 1175-1182 (2005)
Soumya Mukherjee et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(45), 11771-11778 (2018-05-29)
Fluorous organic building blocks were utilized to develop two self-assembled, hydrophobic, fluorinated porous organic polymers (FPOPs), namely, FPOP-100 and FPOP-101. Comprehensive mechanical analyses of these functionalised triazine network polymers marked the introduction of mechanical stiffness among all porous organic network
Robert J Strife et al.
Journal of chromatography. A, 1216(41), 6970-6973 (2009-09-08)
The assessment of human exposure to specific isomers of dimethylanilines (DMA's) is of interest for the evaluation of potential exposure-health outcome relationships. Improved analytical methods will help in identifying the environmental sources of such exposures. The separation of all six
Ming-Wei Chao et al.
Toxicological sciences : an official journal of the Society of Toxicology, 141(1), 300-313 (2014-06-29)
Epidemiological studies have demonstrated extensive human exposure to the monocyclic aromatic amines, particularly to 3,5-dimethylaniline, and found an association between exposure to these compounds and risk for bladder cancer. Little is known about molecular mechanisms that might lead to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service