Skip to Content
Merck
All Photos(1)

Documents

137766

Sigma-Aldrich

Diethyl oxalpropionate

≥95%

Synonym(s):

Diethyl 2-methyl-3-oxosuccinate, Methyloxo-butanedioic acid diethyl ester, NSC 33946

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH(CH3)COCOOC2H5
CAS Number:
Molecular Weight:
202.20
Beilstein:
1783697
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

138 °C/23 mmHg (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)C(=O)C(=O)OCC

InChI

1S/C9H14O5/c1-4-13-8(11)6(3)7(10)9(12)14-5-2/h6H,4-5H2,1-3H3

InChI key

OQOCQBJWOCRPQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Diethyl oxalpropionate was used in the synthesis of 2-hydroxy-3-methylsuccinic acid. It was used as starting reagent in the synthesis of racemic 4-alkyloxycarbonyl-3,3-dimethyl-2-oxetanones and 4-quinolinone 2-carboxylic acid.
Reactant involved in:
  • Synthesis of anticancer and antiviral agents
  • Oxidation by organohypervalent iodine reagent
  • Synthesis of monomers for preparation of functional polyesters
  • Synthesis of calpain inhibitors
  • Preparation of human A2A receptor antagonists
  • Structural studies of dihydropteroate synthase inhibitors

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Hyuk Nam et al.
Bioorganic & medicinal chemistry letters, 18(1), 205-209 (2007-11-21)
Calpains are involved in a variety of calcium-regulated cellular processes, such as signal transduction, cell proliferation, differentiation, and apoptosis. Excessive calpain activation contributes to serious cellular damage and has been reported in many pathological conditions. 4-Quinolinone 2-carboxamide derivatives were prepared
Synthesis of New Homopolyester and Copolyesters by Anionic Ring-opening Polymerization of a, a', ?-Trisubstituted ?-Lactones.
Barbaud C, et al.
Macromolecular Chemistry and Physics, 205(2), 199-207 (2004)
Leanne A Pearson et al.
The Journal of biological chemistry, 282(7), 4681-4692 (2006-12-05)
The cyanobacterium Microcystis aeruginosa is widely known for its production of the potent hepatotoxin microcystin. This cyclic heptapeptide is synthesized non-ribosomally by the thio-template function of a large modular enzyme complex encoded within the 55-kb microcystin synthetase gene (mcy) cluster.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service