Skip to Content
Merck
All Photos(1)

Key Documents

133477

Sigma-Aldrich

1-Amino-2-naphthol hydrochloride

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6OH · HCl
CAS Number:
Molecular Weight:
195.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

crystals

mp

250 °C (dec.) (lit.)

SMILES string

Cl.Nc1c(O)ccc2ccccc12

InChI

1S/C10H9NO.ClH/c11-10-8-4-2-1-3-7(8)5-6-9(10)12;/h1-6,12H,11H2;1H

InChI key

DEKREBCFNLUULC-UHFFFAOYSA-N

General description

1-Amino-2-naphthol undergoes condensation with methylene-substituted azaheterocycles in presence of an oxidizing agent to yield photochromic spirooxazines.

Application

1-Amino-2-naphthol hydrochloride was used in a study on degradation of the azo dye Acid Orange 7 in batch anaerobic unstirred assay.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anita Sztojkov-Ivanov et al.
Journal of separation science, 28(18), 2505-2510 (2006-01-13)
The enantiomers of 1-(alpha-aminobenzyl)-2-naphthol and 2-(alpha-aminobenzyl)-1-naphthol analogs were separated isothermally on a cellulose-tris-3,5-dimethylphenyl carbamate-based chiral stationary phase (Chiralcel OD-H), at 10 degrees C increments in the range of 5-35 degrees C, using n-hexane/2-propanol/diethylamine as mobile phase. The mobile phase composition
Anaerobic treatment of azo dye Acid Orange 7 under batch conditions.
Mendez-Paz D, et al.
Enzyme and Microbial Technology, 36(2), 264-272 (2005)
Synthesis of photochromic spirooxazines from 1-amino-2-naphthols
Lokshin V, et al.
Tetrahedron, 53(28), 9669-9678 (1997)
Yang Mu et al.
Environmental science & technology, 43(13), 5137-5143 (2009-08-14)
Azo dyes are ubiquitously used in the textile industry. These dyes need to be removed from the effluent prior to discharge to sewage due to their intense color and toxicity. In this study we investigated the use of a bioelectrochemical
D Dillon et al.
Mutagenesis, 9(4), 295-299 (1994-07-01)
D & C Red No. 9 is a monoazo dye used for manufacturing printing inks, rubber and plastics, and as an additive in cosmetics and drugs. In an NTP carcinogenicity study in rats and mice it induced splenic sarcomas and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service