Skip to Content
Merck
All Photos(1)

Key Documents

131598

Sigma-Aldrich

Pyrene

crystalline, 95%

Synonym(s):

Benzo[def]phenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
Beilstein:
1307225
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

form

crystalline

mp

145-148 °C (lit.)

SMILES string

c1cc2ccc3cccc4ccc(c1)c2c34

InChI

1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H

InChI key

BBEAQIROQSPTKN-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

435.2 °F

Flash Point(C)

224 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrene fluorescence lifetime as a probe for oxygen penetration of micelles.
M W Geiger et al.
Photochemistry and photobiology, 22(6), 273-276 (1975-12-01)
Photoconductivity of liquid crystalline derivatives of pyrene and carbazole.
Sienkowska, Monika J., et al.
Journal of Materials Chemistry, 17(14), 1392-1398 (2007)
S Fischkoff et al.
The Journal of general physiology, 65(5), 663-676 (1975-05-01)
Quenching of pyrene fluorescence by oxygen was used to determine oxygen diffusion coefficients in phospholipid dispersions and erythrocyte plasma membranes. The fluorescence intensity and lifetime of pyrene in both artificial and natural membranes decreases about 80% in the presence of
Youn Sun Kim et al.
Chemical communications (Cambridge, England), 47(31), 8907-8909 (2011-06-18)
A new pyrene-cored π-conjugated molecule has been synthesized through Sonogashira coupling reaction. The single-crystalline microribbon-based FET exhibited the highest mobility of 0.7 cm(2) V(-1) s(-1) (I(on)/I(off) > 10(6)). Single-crystalline microribbons were employed to operate in an organic phototransistor (OPT) under
Pyrene fluorescence study of chitosan self-association in aqueous solution.
Amiji, M. M.
Carbohydrate Polymers, 26(3), 211-213 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service