Skip to Content
Merck
All Photos(2)

Key Documents

129577

Sigma-Aldrich

Magnesium acetylacetonate dihydrate

98%

Synonym(s):

Magnesium 2,4 pentanedionate dihydrate, Mg(acac)2

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€574.00

€574.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€574.00

About This Item

Linear Formula:
[CH3COCHC(O)CH3]2Mg · 2H2O
CAS Number:
Molecular Weight:
258.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€574.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

core: magnesium

mp

265 °C (dec.) (lit.)

SMILES string

O.O.CC(=O)\C=C(\C)O[Mg]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Mg.2H2O/c2*1-4(6)3-5(2)7;;;/h2*3,6H,1-2H3;;2*1H2/q;;+2;;/p-2/b2*4-3-;;;

InChI key

LDRHDOBLZDBGOP-VGKOASNMSA-L

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service