Recommended Products
Quality Level
Assay
97%
mp
146-148 °C (lit.)
SMILES string
[O-][N+](=O)c1cc([N+]([O-])=O)c2ccccc2c1
InChI
1S/C10H6N2O4/c13-11(14)8-5-7-3-1-2-4-9(7)10(6-8)12(15)16/h1-6H
InChI key
ULALSFRIGPMWRS-UHFFFAOYSA-N
Related Categories
Application
1,3-Dinitronaphthalene was used to study the photocatalytic oxidation reaction of 1,3-dinitronaphthalene in the presence of TiO(2) Degussa (P-25 grade).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Acta crystallographica. Section C, Crystal structure communications, 47 ( Pt 10), 2252-2254 (1991-10-15)
C10H6N2O4, Mr = 218.2, triclinic, P1, a = 7.5552 (8), b = 7.763 (1), c = 9.073 (1) A, alpha = 110.742 (9), beta = 99.570 (9), gamma = 100.309 (9) degrees, V = 474.1 (2) A3, D chi =
Chemosphere, 75(8), 1008-1014 (2009-02-24)
A combination of photocatalytic oxidation experiments and quantum mechanical calculations was used in order to describe the mechanism and the nature of the photocatalytic oxidation reactions of dinitronaphthalane isomers and interprete their reactivities within the framework of the Density Functional
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 28(4), 411-415 (2012-04-14)
The electrochemical behavior and application of a new sensor, a silver solid amalgam paste electrode (AgSA-PE), based on the mixture of a fine silver solid amalgam powder (60:40 (w(Hg)/w(Ag))) and a suitable organic pasting liquid (Paraffin oil) in a ratio
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service