Skip to Content
Merck
All Photos(1)

Key Documents

127485

Sigma-Aldrich

5-Chloro-2-oxindole

98%

Synonym(s):

5-Chlorooxindole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€20.40

€20.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€20.40

About This Item

Empirical Formula (Hill Notation):
C8H6ClNO
CAS Number:
Molecular Weight:
167.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€20.40


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

194-197 °C (lit.)

solubility

methanol: soluble 2.5 mL, clear

functional group

chloro

SMILES string

Clc1ccc2NC(=O)Cc2c1

InChI

1S/C8H6ClNO/c9-6-1-2-7-5(3-6)4-8(11)10-7/h1-3H,4H2,(H,10,11)

InChI key

WWJLCYHYLZZXBE-UHFFFAOYSA-N

General description

5-Chloro-2-oxindole (5-Chlorooxindole) is a starting material for tenidap sodium, a pharmaceutical drug candidate[1].

Application

5-Chloro-2-oxindole was used for the quantitation of 5-chloro-2-oxindole, concomitantly with all of its potential positional isomers using a single, highly specific, normal-phase chromatographic system[1].

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Colgan et al.
Journal of pharmaceutical and biomedical analysis, 14(7), 825-833 (1996-05-01)
5-Chlorooxindole (5-CO) is a starting material for tenidap sodium, a pharmaceutical drug candidate produced by Pfizer. To insure potency and purity of the drug substance, it is necessary to demonstrate that commercial supplies of 5-CO are free from elevated levels
Boris Letribot et al.
Molecules (Basel, Switzerland), 23(6) (2018-06-13)
Alkylidene oxindoles are important functional moieties and building blocks in pharmaceutical and synthetic chemistry. Our interest in biologically active compounds focused our studies on the synthesis of novel oxindoles, bearing on the exocyclic double bond at the C8, CN, and
Ayman M Saleh et al.
Molecules (Basel, Switzerland), 19(9), 13076-13092 (2014-08-27)
A selected set of substituted pyridone-annelated isoindigos 3a-f has been synthesized via interaction of 5- and 6-substituted oxindoles 2a-f with 6-ethyl-1,2,9-trioxopyrrolo[3,2-f]quinoline-8-carboxylic acid (1) in acetic acid at reflux. Among these isoindigos, the 5'-chloro and 5'-bromo derivatives 3b and 3d show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service