Skip to Content
Merck
All Photos(1)

Key Documents

127345

Sigma-Aldrich

4-(2-Thiazolylazo)resorcinol

97%

Synonym(s):

2-(2,4-Dihydroxyphenylazo)thiazole, TAR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N3O2S
CAS Number:
Molecular Weight:
221.24
Beilstein:
204257
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

97%

mp

211 °C (dec.) (lit.)

SMILES string

Oc1ccc(\N=N\c2nccs2)c(O)c1

InChI

1S/C9H7N3O2S/c13-6-1-2-7(8(14)5-6)11-12-9-10-3-4-15-9/h1-5,13-14H/b12-11+

InChI key

SHNIKUXMZFPPCS-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful for the spectrophotometric determination of trace metals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Benvidi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(5), 1380-1385 (2011-03-01)
The acidity constants of 4-(2-thiazolylazo)-resorcinol (TAR) were determined by the principal component-wavelet neural network (WNN). Biprotic acid mass balance equations, the distribution functions and the corresponding spectral profiles which were generated by a Gaussian model, have been considered to simulate
S R Bergmann et al.
Clinica chimica acta; international journal of clinical chemistry, 104(1), 53-63 (1980-05-21)
A simple and rapid colorimetric assay for non-esterified fatty acids (NEFA) was developed employing extraction of samples with 0.1 mol/l glycine (pH 2.7) and methoxyethanol : butyl ether, and formation of a copper-fatty acid soap detected with 4-(2-thiazolylazo)-resorcinol. The method
Yue-Feng Zhou et al.
The Analyst, 136(2), 282-284 (2010-11-11)
A simple cyclopalladated complex of 4-(2-thiazolylazo)resorcinol showed a specific red-to-green colour change upon addition of organotin species in the acetonitrile-water medium.
Mustafa Soylak et al.
Journal of hazardous materials, 182(1-3), 704-709 (2010-07-17)
A sensitive and simple separation-enrichment technique for the determination of trace amounts of some metal ions was described. By the passage of aqueous samples including Fe(III), Cu(II), Ni(II) and Co(II) ions-4-(2-thiazolylazo) resorcinol (TAR) chelates through Diaion SP-850 column, metal chelates
Y Kalyan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(5), 1235-1241 (2009-11-03)
A membrane optode was developed utilizing the 8-hydroxyquinaldine (HQ) facilitated preconcentration of UO(2)(2+) ions and subsequent colored complex formation of UO(2)(2+) with 4-(2-thiazolylazo)-resorcinol (TAR) in optode matrix. The composition of the membrane optode was optimized by scanning several extractants immobilized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service