Skip to Content
Merck
All Photos(2)

Key Documents

126411

Sigma-Aldrich

1,3-Dimethylbutylamine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€59.60

€59.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€59.60

About This Item

Linear Formula:
(CH3)2CHCH2CH(CH3)NH2
CAS Number:
Molecular Weight:
101.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€59.60


Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.4085 (lit.)

bp

108-110 °C (lit.)

density

0.717 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC(C)CC(C)N

InChI

1S/C6H15N/c1-5(2)4-6(3)7/h5-6H,4,7H2,1-3H3

InChI key

UNBMPKNTYKDYCG-UHFFFAOYSA-N

Application

1,3-Dimethylbutylamine has been used to study the application of unfunctionized polymethacrylate resin (TSKgel G3000PWXL) as a stationary phase in liquid chromatography with UV detection[1].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazutoku Ohta et al.
Journal of chromatography. A, 1039(1-2), 161-169 (2004-07-15)
The application of unfunctionized polymethacrylate resin (TSKgel G3000PWXL) as a stationary phase in liquid chromatography with UV detection for C1-C7 aliphatic monocarboxylic acids (formic acid, acetic acid, propionic acid, butyric acid, isovaleric acid, valeric acid, 3,3-dimethylbutyric acid, 4-methylvaleric acid, hexanoic
Hong Ren et al.
Journal of biotechnology, 241, 33-41 (2016-11-14)
Asymmetric synthesis of chiral amines by amine dehydrogenases (AmDH), which catalyzed reductive amination of ketones with high enantioselectivity, is an ideal route for production of chiral amine. In this study, a facile approach is proposed to immobilize unstable amine dehydrogenase
Anna Rickli et al.
European journal of pharmacology, 859, 172515-172515 (2019-07-03)
Preworkout supplements ("boosters") are used to enhance physical and mental performance during workouts. These products may contain various chemical substances with undefined pharmacological activity. We investigated whether substances that are contained in commercially available athletic multiple-ingredient preworkout supplements exert amphetamine-type
Pieter A Cohen et al.
Clinical toxicology (Philadelphia, Pa.), 56(6), 421-426 (2017-11-09)
The United States Food and Drug Administration banned the stimulant 1,3-dimethylamylamine (1,3-DMAA) from dietary supplements and warned consumers that the stimulant can pose cardiovascular risks ranging from high blood pressure to heart attacks. We designed our study to determine if

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service