Skip to Content
Merck
All Photos(1)

Key Documents

125571

Sigma-Aldrich

5-Chloro-2-mercaptobenzothiazole

technical grade, ≥90%

Synonym(s):

5-Chloro-2-benzothiazolethiol, CMBT

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€131.00

€131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€131.00

About This Item

Empirical Formula (Hill Notation):
C7H4ClNS2
CAS Number:
Molecular Weight:
201.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€131.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

≥90%

mp

198-200 °C (lit.)

functional group

chloro

SMILES string

Sc1nc2cc(Cl)ccc2s1

InChI

1S/C7H4ClNS2/c8-4-1-2-6-5(3-4)9-7(10)11-6/h1-3H,(H,9,10)

InChI key

NKYDKCVZNMNZCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Chloro-2-mercaptobenzothiazole(CMBZT) reacts with iodine in a molar ratio of 1:1 to form charge-transfer complex (CMBZT)I2[1].

Application

5-Chloro-2-mercaptobenzothiazole (CMBZT) has been used in the analysis of lipopolysaccharide by negative-ion matrix-assisted laser desorption ionization-time of flight mass spectrometry[2]. It has been used in the synthesis of the new mixed ligand silver(I) chloride complex with the heterocyclic thioamide CMBZT and tri(p-tolyl)phosphine[3]. Found to be more sensitive than DHB for analysis of high mannose N-linked glycans. Used in analysis of peptidoglycan muropeptides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adeline M Hajjar et al.
Infection and immunity, 74(12), 6730-6738 (2006-09-20)
Francisella tularensis is an intracellular gram-negative bacterium that is highly infectious and potentially lethal. Several subspecies exist of varying pathogenicity. Infection by only a few organisms is sufficient to cause disease depending on the model system. Lipopolysaccharide (LPS) of gram-negative
Chen Chen et al.
Biotechnology for biofuels, 11, 155-155 (2018-07-12)
Polysaccharide monooxygenases (PMOs) of the auxiliary activity 9 (AA9) family have been reported to oxidize C1, C4, and C6 positions in cellulose. However, currently no direct evidence exists that PMOs oxidize C6 positions in cellulose, and molecular mechanism of C1, C4
Natasha A Bencivenga-Barry et al.
Journal of bacteriology, 202(3) (2019-11-13)
Bacteroides is one of the most prominent genera in the human gut microbiome, and study of this bacterial group provides insights into gut microbial ecology and pathogenesis. In this report, we introduce a negative selection system for rapid and efficient
Synthesis and Characterization of (PTU) I2 (PTU= 6-n-propyl-2-thiouracil) and (CMBZT) I2 (CMBZT= 5-chloro-2-mercaptobenzothiazole) and Possible Implications for the Mechanism of Action of Anti-Thyroid Drugs.
Antoniadis CD, et al.
European Journal of Inorganic Chemistry, 2003(8), 1635-1640 (2003)
L Kyros et al.
Bioinorganic chemistry and applications, 386860-386860 (2010-04-10)
The synthesis and characterization of the silver(I) chloride complex of formula {[AgCI(CMBZT)(TPTP)(2)] . (MeOH)} (1) (CMBZT = 5-chloro-2-mercaptobenzothiazole, TPTP = tris(p-tolyl)phosphine) is described. Also the structure of the hydrate derivative {[AgCI(TPTP)(3)] . (0.5 . H(2)O)} (2) of the corresponding known

Protocols

Explore mass spectrometry analysis of glycans for glycomic & glycoproteomic neutral & acidic glycan analysis. See a general mass spec glycan analysis procedure.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service