Skip to Content
Merck
All Photos(1)

Key Documents

12234

Sigma-Aldrich

2-(4-Boc-piperazino)-2-(2-fluorophenyl)acetic acid

≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€148.00
1 G
€475.00

€148.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€148.00
1 G
€475.00

About This Item

Empirical Formula (Hill Notation):
C17H23FN2O4
CAS Number:
Molecular Weight:
338.37
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€148.00


Estimated to ship on24 April 2025


Request a Bulk Order

Product Name

2-(4-Boc-piperazino)-2-(2-fluorophenyl)acetic acid, purum, ≥97.0% (HPLC)

grade

purum

Quality Level

Assay

≥97.0% (HPLC)

form

solid

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCN(CC1)C(C(O)=O)c2ccccc2F

InChI

1S/C17H23FN2O4/c1-17(2,3)24-16(23)20-10-8-19(9-11-20)14(15(21)22)12-6-4-5-7-13(12)18/h4-7,14H,8-11H2,1-3H3,(H,21,22)

InChI key

ZSGKIKRNLJANGA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service