Skip to Content
Merck
All Photos(1)

Documents

119482

Sigma-Aldrich

8-Bromoguanosine hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12BrN5O5 · xH2O
CAS Number:
Molecular Weight:
362.14 (anhydrous basis)
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −28.8°, c = 2 in DMSO/H2O, 1:1

mp

222 °C (dec.) (lit.)

SMILES string

O.NC1=Nc2c(nc(Br)n2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingyuan Jiao et al.
Journal of separation science, 37(21), 3033-3044 (2014-08-20)
We first detected aberrant nucleoside levels in the plasma, urine, bile, and tissues from cases and controls to explore them as biomarkers in the diagnosis of gallbladder cancer. Reversed-phase high-performance liquid chromatography was used to assess the levels of ten

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service