Skip to Content
Merck
All Photos(1)

Documents

119474

Sigma-Aldrich

N-Hydroxyurethane

Synonym(s):

N-Carbethoxyhydroxylamine, Ethyl N-hydroxycarbamate, Hydroxycarbamic acid ethyl ester, NSC 71045, NSC 83629

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HONHCOOCH2CH3
CAS Number:
Molecular Weight:
105.09
Beilstein:
1747529
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.445 (lit.)

bp

113-116 °C/3 mmHg (lit.)

storage temp.

−20°C

SMILES string

CCOC(=O)NO

InChI

1S/C3H7NO3/c1-2-7-3(5)4-6/h6H,2H2,1H3,(H,4,5)

InChI key

VGEWEGHHYWGXGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Hydroxyurethane was used to synthesize N-methyl-O-benzylhydroxylamine and N-isopropyl-O-methylhydroxylamine.
Reactant involved in:
  • Synthesis of molecules used for intermolecular Sharpless aminohydroxylation reactions
  • Intermolecular ortho-C-H amidation of anilides
  • Cinchona alkaloid-catalyzed asymmetric cycloaddition
  • Allylic arylation

Biochem/physiol Actions

N-Hydroxyurethane causes the chromosomal fragmentation at millimolar concentrations and cell toxicity in cultured normal human leukocytes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Nomura et al.
Cancer research, 43(11), 5156-5162 (1983-11-01)
To learn the effects of tumor inhibitors on chemically induced malformations, caffeine, antipain, and 13-trans-retinoic acid were given to pregnant ICR/Jcl mice after a single dose of urethan, N-hydroxyurethan, N-methyl-N-nitrosourea, N-ethyl-N-nitrosourea, or 4-nitroquinoline 1-oxide, which induces about 50% of the
Comparative carcinogenicities and mutagenicities of vinyl carbamate, ethyl carbamate, and ethyl N-hydroxycarbamate.
G A Dahl et al.
Cancer research, 40(4), 1194-1203 (1980-04-01)
K Sugihara et al.
Journal of pharmacobio-dynamics, 6(9), 677-683 (1983-09-01)
The present study provides the evidence that liver aldehyde oxidase in the presence of its electron donors can catalyze the reduction of N-hydroxyurethane to urethane under anaerobic conditions. Guinea pig liver 9000 X g supernatant and cytosol, but not liver
Kinetic applications of electron paramagnetic resonance spectroscopy. 28. N-Alkoxy-N-alkylamino, N-alkoxyamino, and N-alkoxyanilino radicals.
Kaba RA and Ingold KU
Journal of the American Chemical Society, 98(23), 7375-7380 (1976)
P M Weiss et al.
Biochemistry, 23(19), 4346-4350 (1984-09-11)
The true substrate for the pyruvate kinase catalyzed phosphorylation of hydroxylamine at high pH which is activated by bicarbonate is shown to be N-hydroxycarbamate, since a lag is seen when the reaction is started by the addition of bicarbonate or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service