Skip to Content
Merck
All Photos(2)

Documents

116157

Sigma-Aldrich

1,4-Dibromo-2,5-dimethylbenzene

98%

Synonym(s):

2,5-Dibromo-p-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H2Br2
CAS Number:
Molecular Weight:
263.96
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

261 °C (lit.)

mp

72-74 °C (lit.)

SMILES string

Cc1cc(Br)c(C)cc1Br

InChI

1S/C8H8Br2/c1-5-3-8(10)6(2)4-7(5)9/h3-4H,1-2H3

InChI key

QENIALCDPFDFHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,4-dibromo-2,5-dimethylbenzene has been used in the preparation of 4,4″-diformyl-2′,5′-dimethyl-1,1′.4′,1″-terphenyl. It has also been used in the preparation of 1,4-diformyl-2,5-dimethylbenzene.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuxiang Du et al.
Light, science & applications, 9, 151-151 (2020-09-10)
Tuneable microlasers that span the full visible spectrum, particularly red, green, and blue (RGB) colors, are of crucial importance for various optical devices. However, RGB microlasers usually operate in multimode because the mode selection strategy cannot be applied to the
Nikolai Kuhnert et al.
Organic & biomolecular chemistry, 1(7), 1157-1170 (2003-08-21)
The synthesis of aromatic dicarboxaldehydes, using dilithiation methodology is described along with their reactivity, in the [3 + 3] cyclocondensation reaction, with (1R,2R)-diaminocyclohexane to give trianglimine macrocycles. The scope and limitations of the cyclocondensation reaction are studied and some comments
Nikolai Kuhnert et al.
Organic & biomolecular chemistry, 3(10), 1911-1921 (2005-05-13)
The synthesis of aromatic dicarboxaldehydes is described along with their reactivity in the [3 + 3] cyclocondensation reaction with (1R,2R)-diaminocyclohexane to give trianglimine macrocycles. In particular, the scope and limitation of the reaction with regard to complete control of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service