116025
Mandelonitrile
technical grade
Synonym(s):
α-Hydroxyphenylacetonitrile, Benzaldehyde cyanohydrin, Mandelic acid nitrile
About This Item
Recommended Products
grade
technical grade
refractive index
n20/D 1.530 (lit.)
bp
170 °C (lit.)
solubility
alcohol: freely soluble
chloroform: freely soluble
diethyl ether: freely soluble
density
1.117 g/mL at 25 °C (lit.)
SMILES string
OC(C#N)c1ccccc1
InChI
1S/C8H7NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H
InChI key
NNICRUQPODTGRU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
206.6 °F - closed cup
Flash Point(C)
97 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service