Skip to Content
Merck
All Photos(1)

Key Documents

115924

Sigma-Aldrich

3-Methyl-1-butanethiol

97%

Synonym(s):

Isoamyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2SH
CAS Number:
Molecular Weight:
104.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

vapor pressure

41.4 mmHg ( 37.7 °C)

Assay

97%

refractive index

n20/D 1.4432 (lit.)

bp

117-118 °C (lit.)

density

0.835 g/mL at 25 °C (lit.)

functional group

thiol

SMILES string

CC(C)CCS

InChI

1S/C5H12S/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

GIJGXNFNUUFEGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-1-butanethiol is one of the volatile components obtained by the GC-MS analysis of the anal sac secretion from the spotted skunk, Spilogale putorius[1].

Application

3-Methyl-1-butanethiol has been used as an organic clapping layer in the synthesis of monolayer-capped 5 nm gold nanoparticles[2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W F Wood et al.
Journal of chemical ecology, 17(7), 1415-1420 (1991-07-01)
GC-MS analysis of the anal sac secretion from the spotted skunk,Spilogale putorius, showed three major volatile components: (E)-2-butene-1-thiol, 3-methyl-1-butanethiol, and 2-phenylethanethiol. Minor volatile components identified from this secretion were: phenylmethanethiol, 2-methylquinoline, 2-quinolinemethanethiol, bis[(E)-2-butenyl] disulfide, (E)-2-butenyl 3-methylbufyl disulfide, bis(3-methylbutyl) disulfide. All
G Peng et al.
British journal of cancer, 103(4), 542-551 (2010-07-22)
Tumour growth is accompanied by gene and/or protein changes that may lead to peroxidation of the cell membrane species and, hence, to the emission of volatile organic compounds (VOCs). In this study, we investigated the ability of a nanosensor array

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service