Skip to Content
Merck
All Photos(2)

Key Documents

114596

Sigma-Aldrich

N,N-Diphenylhydrazine hydrochloride

97%

Synonym(s):

N,N-Diphenylhydrazine hydrochloride (asym.)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€121.00

€121.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€121.00

About This Item

Linear Formula:
(C6H5)2NNH2 · HCl
CAS Number:
Molecular Weight:
220.70
Beilstein:
3569340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€121.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

SMILES string

Cl.NN(c1ccccc1)c2ccccc2

InChI

1S/C12H12N2.ClH/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10H,13H2;1H

InChI key

MIVUDWFNUOXEJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Diphenylhydrazine hydrochloride is used in the synthesis of 2-Vinyloxyethyloxy-4-diethylaminophenyl-1-carbaldehyde N,N-diphenylhydrazone (hole-transporting hydrazine)[1].

Other Notes

Small amounts of moisture cause melting point to vary greatly

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduction of N-nitrosodiphenylamine to the corresponding hydrazine by guinea pig liver preparations.
K Tatsumi et al.
Chemical & pharmaceutical bulletin, 30(10), 3842-3845 (1982-10-01)
K Tatsumi et al.
Biochemical and biophysical research communications, 118(3), 958-963 (1984-02-14)
The present study provides the first evidence for in vitro metabolic conversion of a 1,1-disubstituted hydrazine to the corresponding nitrosamine. The study shows that superoxide radical which is generated by NADPH-cytochrome c reductase is involved in the oxidation of 1,1-diphenylhydrazine
New reactive hole-transporting hydrazone and its adducts with diol and dithiol.
Journal of Optoelectronics and Advanced Materials, 8(4), 1533-1533 (2006)
João M S Cardoso et al.
Journal of inorganic biochemistry, 166, 55-63 (2016-11-12)
Camphorsulphonylimine complexes [Ag(NO
Sara S M Fernandes et al.
ACS omega, 3(10), 12893-12904 (2018-11-10)
A series of push-pull heterocyclic N,N-diphenylhydrazones were prepared to study the effect of structural modifications (different π-spacers and electron-withdrawing groups) on the optical (linear and nonlinear) and electronic properties of the molecules. The photovoltaic response of dye-sensitized solar cells assembled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service