Skip to Content
Merck
All Photos(2)

Key Documents

114278

Sigma-Aldrich

(2-Chloroethoxy)benzene

98%

Synonym(s):

β-Chlorophenetole, 2-Chloroethyl phenyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€58.90

€58.90


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 ML
€58.90

About This Item

Linear Formula:
C6H5OCH2CH2Cl
CAS Number:
Molecular Weight:
156.61
Beilstein:
971048
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€58.90


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

98%

refractive index

n20/D 1.534 (lit.)

bp

97-98 °C/15 mmHg (lit.)

density

1.129 g/mL at 25 °C (lit.)

functional group

chloro
phenoxy

SMILES string

ClCCOc1ccccc1

InChI

1S/C8H9ClO/c9-6-7-10-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

VQUYNUJARXBNPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(2-Chloroethoxy)benzene is used in the coupling reaction of benzaldehyde, cinnamyltrimethylsilane and anisole in the synthesis of pseudo-symmetrical tamoxifen derivatives[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isamu Shiina et al.
Bioorganic & medicinal chemistry letters, 17(9), 2421-2424 (2007-03-10)
Three new pseudo-symmetrical tamoxifen derivatives, RID-B (15), C (16), and D (17), were synthesized via the novel three-component coupling reaction, and the structure-activity relationships of the pseudo-symmetrical tamoxifen derivatives were examined. It was discovered that 15 strongly inhibits the viability

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service