Skip to Content
Merck
All Photos(2)

Documents

114162

Sigma-Aldrich

Chlorotriphenylsilane

96%

Synonym(s):

TPSCl, Triphenylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SiCl
CAS Number:
Molecular Weight:
294.85
Beilstein:
1820487
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

91-94 °C (lit.)

SMILES string

Cl[Si](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15ClSi/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

MNKYQPOFRKPUAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Okamoto
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 112(6), 409-413 (1992-06-01)
The retention behavior and selectivity of 30 kinds of phenyl-modified porous glasses and silicas, prepared from solutions of phenyldimethylchlorosilane, diphenylmethylchlorosilane or triphenylchlorosilane in xylene, were studied by high-performance liquid chromatography using carbamazepine and diphenylhydantoin as model compounds. From the elemental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service