Skip to Content
Merck
All Photos(1)

Documents

113964

Sigma-Aldrich

2-Bromo-4,6-dinitroaniline

94%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(NO2)2NH2
CAS Number:
Molecular Weight:
262.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

94%

form

solid

mp

151-153 °C (lit.)

SMILES string

Nc1c(Br)cc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4BrN3O4/c7-4-1-3(9(11)12)2-5(6(4)8)10(13)14/h1-2H,8H2

InChI key

KWMDHCLJYMVBNS-UHFFFAOYSA-N

General description

2-Bromo-4,6-dinitroaniline is an important metabolite of the azo dye Disperse Blue 79. It can be easily recognized by all rat monoclonal antibodies.

Application

2-Bromo-4,6-dinitroaniline was used as a model chemical in the inter-laboratory evaluation of the bioluminescent Salmonella reverse mutation assay for detecting mutagenicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Peng et al.
Environmental science & technology, 50(23), 12669-12677 (2016-12-10)
Characterization of toxicological profiles by use of traditional targeted strategies might underestimate the risk of environmental mixtures. Unbiased identification of prioritized compounds provides a promising strategy for meeting regulatory needs. In this study, untargeted screening of brominated compounds in house
H M Chopade et al.
Journal of toxicology and environmental health, 17(1), 37-50 (1986-01-01)
The disposition of [14C]-2-bromo-4,6-dinitroaniline (BDNA) was studied in male F344 rats following oral or intravenous (iv) administration. The gastrointestinal absorption of BDNA was nearly complete and was not affected by dose in the range (10-100 mumol/kg body weight) studied. Following
Joel Ackerman et al.
Mutagenesis, 24(5), 433-438 (2009-07-08)
We have developed the bioluminescent Salmonella reverse mutation assay as a tool for detecting mutagenicity applicable for high-throughput screening of new chemicals. In this study, we report the inter-laboratory evaluation of the assay using 10 model chemicals in five independent
Petra M Krämer et al.
Analytical and bioanalytical chemistry, 391(5), 1821-1835 (2008-05-28)
The development and characterization of one rat monoclonal antibody (mAb) for 2,4-dinitroaniline and of two rat mAbs for 2,6-dinitroaniline are described. With the immunization of rats with 2,4,6-trinitrophenyl-glycylglycine-keyhole limpet hemocyanine (KLH) conjugate one mAb (PK 5H6) has been developed and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service