Skip to Content
Merck
All Photos(1)

Key Documents

11214

Sigma-Aldrich

L-Aspartic acid 4-tert-butyl ester

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO4
CAS Number:
Molecular Weight:
189.21
Beilstein:
4671089
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

L-Aspartic acid 4-tert-butyl ester, ≥98.0% (TLC)

Assay

≥98.0% (TLC)

optical activity

[α]20/D −9±2°, c = 2% in methanol: water (4:1)

reaction suitability

reaction type: solution phase peptide synthesis

color

white

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)C[C@H](N)C(O)=O

InChI

1S/C8H15NO4/c1-8(2,3)13-6(10)4-5(9)7(11)12/h5H,4,9H2,1-3H3,(H,11,12)/t5-/m0/s1

InChI key

MXWMFBYWXMXRPD-YFKPBYRVSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service