Skip to Content
Merck
All Photos(1)

Key Documents

10924

Sigma-Aldrich

Sodium cyclohexanesulfonate

≥98.0% (T)

Synonym(s):

Cyclohexanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NaO3S
CAS Number:
Molecular Weight:
186.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

solid

impurities

≤1% water

SMILES string

[Na+].[O-]S(=O)(=O)C1CCCCC1

InChI

1S/C6H12O3S.Na/c7-10(8,9)6-4-2-1-3-5-6;/h6H,1-5H2,(H,7,8,9);/q;+1/p-1

InChI key

DIIKAKPJAGLSOD-UHFFFAOYSA-M

General description

Sodium cyclohexanesulfonate is a white to faintly beige powder to fine crystals with lump.

Application

Sodium cyclohexanesulfonate was used to study the influence of various hydrophobic ionic excipients on viscosity of concentrated solutions of therapeutic proteins. It was used as an ionic hydrophobic excipient that substantially decrease the viscosity of concentrated (300-400 mg/mL) aqueous solutions of bovine serum albumin and γ-globulin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Du et al.
Biotechnology and bioengineering, 108(3), 632-636 (2011-01-20)
Reducing viscosities of concentrated solutions of therapeutic proteins is important for their subcutaneous and intravenous delivery. Although inorganic salts and optimizing the pH were previously reported to dramatically lower the viscosity of a monoclonal antibody solution, herein we have determined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service