Skip to Content
Merck
All Photos(1)

Key Documents

108367

Sigma-Aldrich

Cyclobutyl phenyl ketone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC4H7
CAS Number:
Molecular Weight:
160.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

refractive index

n20/D 1.547 (lit.)

bp

114-118 °C/7 mmHg (lit.)

density

1.05 g/mL at 25 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C(C1CCC1)c2ccccc2

InChI

1S/C11H12O/c12-11(10-7-4-8-10)9-5-2-1-3-6-9/h1-3,5-6,10H,4,7-8H2

InChI key

MVEBDOSCXOQNAR-UHFFFAOYSA-N

General description

Cyclobutyl phenyl ketone reacts with 2-butanol and di-t-butyl peroxide to yield valerophenone[1].

Application

Cyclobutyl phenyl ketone was used to study thermal reactions of parent and phenyl substituted N-acyl cyclobutylimines[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some Reactions of Cyclobutylcarbinyl Radical Intermediates.
Neckers DC, et al.
The Journal of Organic Chemistry, 31(2), 622-624 (1966)
Thermal Reactions of N-Acyl Phenylcyclobutylimines.
Wu PL.
J. Chin. Chem. Soc., 45, 183-188 (1998)
Jose Serrano et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(2), 192-208 (2019-03-20)
1. Cyclic phenones are chemicals of interest to the USEPA due to their potential for endocrine disruption to aquatic and terrestrial species.2. Prior to this report, there was very limited information addressing metabolism of cyclic phenones by fish species and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service