Skip to Content
Merck
All Photos(2)

Documents

106887

Sigma-Aldrich

Ferrocenecarboxylic acid

97%

Synonym(s):

(Carboxycyclopentadienyl)cyclopentadienyliron, Carboxyferrocene, Carboxylferrocene, Cyclopentadienecarboxylic acid, Ferrocenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10FeO2
CAS Number:
Molecular Weight:
230.04
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: iron
reagent type: catalyst

mp

210 °C (dec.) (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.OC(=O)[C]2[CH][CH][CH][CH]2

InChI

1S/C6H5O2.C5H5.Fe/c7-6(8)5-3-1-2-4-5;1-2-4-5-3-1;/h1-4H,(H,7,8);1-5H;

InChI key

VUJLGCHOGQEAED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Papanicolaou is a widely used stain in cytopathology. It is a polychrome stain that has the ability to stain several constituents of cell differentially due to the presence of multiple dyes. Papanicolaou stain consists of a basic dye, haematoxylin that stains the nucleus and three acid dyes like light green, eosin and orange G that stain the cytoplasm.

Application

Used to esterify complex mixtures of phenols and alcohols for analysis by GCMS.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewa Nazaruk et al.
Bioelectrochemistry (Amsterdam, Netherlands), 71(1), 8-14 (2007-02-10)
The monoolein-based liquid crystalline cubic phase was used as the matrix to incorporate redox enzymes--glucose (GOx), pyranose (PyOx) oxidases and laccase. Thin layer of the cubic phase embedding GOx or PyOx activated glucose oxidation in the presence and absence of
Shaoling Song et al.
The journal of physical chemistry. B, 114(35), 11689-11695 (2010-08-17)
Poly(4-vinylpyridine) (P4VP) films were electropolymerized on a pyrolytic graphite (PG) electrode surface. The cyclic voltammetric (CV) response of ferrocenedicarboxylic acid (Fc(COOH)(2)) at P4VP film electrodes was very sensitive to the pH and perchlorate (ClO(4)(-)) concentration in testing solutions. Fc(COOH)(2) was
Decoration of cowpea mosaic virus with multiple, redox-active, organometallic complexes.
Nicole F Steinmetz et al.
Small (Weinheim an der Bergstrasse, Germany), 2(4), 530-533 (2006-12-29)
Georg Diehl et al.
Journal of chromatography. A, 974(1-2), 103-109 (2002-12-03)
Rapid liquid chromatographic separations of ferrocenecarboxylic esters of various alcohols and phenols have been achieved on reversed-phase columns of 20 mm length. After separation, the ferrocene derivatives are oxidized electrochemically under formation of the charged ferrocinium species, which are easily
Cedric L Hugelshofer et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(42), 15125-15136 (2016-09-08)
We present a full account of the development of a strategy that culminated in the first total syntheses of the unique oxetane-containing natural product (+)-dictyoxetane and the macrocyclic diterpene (+)-dolabellane V. Our retrosynthetic planning was guided by both classical and nonconventional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service