Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
The journal of physical chemistry. B, 115(30), 9473-9490 (2011-06-21)
Electrostatic properties of proteins are crucial for their functionality. Carboxyamides are small polar groups that, as peptide bonds, are principal structural components of proteins that govern their electrostatic properties. We investigated the medium dependence of the molar polarization and of
This study examines the performance of bench-scale Upflow Anaerobic Sludge Blanket (UASB) reactor in treating simulated wastewaters containing 1,1,2,2-tetrachloroethane (TeCA). Reactors R1 (control) and R2 (containing TeCA) were operated at Hydraulic Retention Times (HRTs) of 36, 30, 24, 18 and
Journal of chromatography. A, 1216(45), 7846-7850 (2009-10-07)
A novel sample preparation method "Dispersive liquid-liquid-liquid microextraction" (DLLLME) was developed in this study. DLLLME was combined with liquid chromatography system to determine chlorophenoxy acid herbicide in aqueous samples. DLLLME is a rapid and environmentally friendly sample pretreatment method. In
Chemical communications (Cambridge, England), 47(31), 8769-8771 (2011-05-28)
Directional Cl···Cl type I and II interactions govern the low-density aggregation of 1,1,2,2-tetrachloroethane molecules in synclinal conformation in the crystalline state at low temperature, whereas the dense molecular packing in high-pressure is achieved for the antiperiplanar conformers and electrostatically less
Transformation of 1,1,2,2-tetrachloroethane (1,1,2,2-TeCA) by Fe(II) in 10% cement slurries was characterized using a batch reactor system. 1,1,2,2-TeCA was completely converted to trichloroethylene (TCE) within 1h in all experiments, even in controls with cement that did not include Fe(II). Therefore