Skip to Content
Merck
All Photos(1)

Key Documents

104825

Sigma-Aldrich

Butyltrichlorosilane

99%

Synonym(s):

Trichlorobutylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SiCl3
CAS Number:
Molecular Weight:
191.56
Beilstein:
1736032
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

149 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

CCCC[Si](Cl)(Cl)Cl

InChI

1S/C4H9Cl3Si/c1-2-3-4-8(5,6)7/h2-4H2,1H3

InChI key

FQEKAFQSVPLXON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyltrichlorosilane (BTS) is a silane based surface modifier that forms a self-assembled monolayer(SAM) by forming a thin outer layer coated on the surface of a variety of metals and metal oxides.[1][2]

Application

BTS can be coated on the glass substrate to introduce the surface alkyl, which can bind well with 2-methacryloyloxyethyl-4-azidobenzoate (MPAz) to convert hydrophophobic glass surfaces to superhydrophilic surfaces.[3] It can functionalize zinc peroxide nanoparticle/polystyrene sulfonate (PSS) hybrid based sensors to improve the limit of detection by three-folds.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface modification of chlorine-passivated silicon nanocrystals
Wang R, et al.
Physical Chemistry Chemical Physics, 15(6), 1815-1820 (2013)
From Si and C encapsulated SiO2 to SiC: exploring the influence of sol-gel polymer substitution on thermally induced nanocrystal formation.
Dasog, Mita, Christina Rachinsky, and Jonathan GC Veinot
Journal of Materials Chemistry, 21(33), 12422-12427 (2011)
Monolayer-controlled deposition of silicon oxide films on gold, silicon, and mica substrates by room-temperature adsorption and oxidation of alkylsiloxane monolayers
Vallant, T., et al.
The Journal of Physical Chemistry B, 104(22), 5309-5317 (2000)
Synthesis of photoreactive phospholipid polymers for use in versatile surface modification of various materials to obtain extreme wettability.
Fukazawa K and Ishihara K
ACS Applied Materials & Interfaces, 5(15), 6832-6836 (2013)
ZnO2 nanohybrid thin film sensor for the detection of ethanol vapour at room temperature using reflectometric interference spectroscopy.
Sebok D and Imre D
Sensors and Actuators B, Chemical, 206(6), 435-442 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service