Skip to Content
Merck
All Photos(1)

Documents

104590

Sigma-Aldrich

N,N′-(o-Phenylene)dimaleimide

99%

Synonym(s):

1,2-Dimaleimidobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8N2O4
CAS Number:
Molecular Weight:
268.22
Beilstein:
249426
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

245-247 °C (dec.) (lit.)

SMILES string

O=C1C=CC(=O)N1c2ccccc2N3C(=O)C=CC3=O

InChI

1S/C14H8N2O4/c17-11-5-6-12(18)15(11)9-3-1-2-4-10(9)16-13(19)7-8-14(16)20/h1-8H

InChI key

UFFVWIGGYXLXPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Imagawa et al.
Journal of biochemistry, 96(6), 1727-1735 (1984-12-01)
A method is described for the preparation of a monomeric Fab'-beta-D-galactosidase conjugate, which is required for the development of a sensitive immunoenzymometric assay. Anti-human IgG F(ab')2 was labeled with 2,4-dinitrophenyl groups, split into Fab' by reduction and reacted with excess
T Chen et al.
Biochemistry, 24(20), 5620-5625 (1985-09-24)
Chemical cross-linking of actin to the 20K and 50K fragments of tryptically cleaved myosin subfragment 1 (S-1) by the zero-length cross-linking reagent 1-ethyl-3-[3-dimethylamino)propyl]carbodiimide (EDC) was used as a probe of the acto-S-1 interface in the presence of nucleotides. The course
Rémi Fronzes et al.
Biochemistry, 42(41), 12038-12049 (2003-10-15)
Subunit h, a 92-residue-long, hydrophilic, acidic protein, is a component of the yeast mitochondrial F1Fo ATP synthase. This subunit, homologous to the mammalian factor F6, is essential for the correct assembly and/or functioning of this enzyme since yeast cells lacking
[Enzymoimmunologic method of determining biologically active substances and its use].
A Szewczuk
Postepy higieny i medycyny doswiadczalnej, 38(5-6), 667-679 (1984-09-01)
T Katoh et al.
European journal of biochemistry, 227(1-2), 459-465 (1995-01-15)
Porcine aorta myosin was reacted with a bifunctional cross-linking reagent, N,N'-o-phenylenedimaleimide. The 17-kDa essential light chain (LC17) in each myosin head was intramolecularly cross-linked within a single myosin molecule. The 34-kDa cross-linked LC17 dimer was isolated and its peptide map

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service