Skip to Content
Merck
All Photos(1)

Key Documents

101869

Sigma-Aldrich

Cycloundecanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H20(=O)
CAS Number:
Molecular Weight:
168.28
Beilstein:
1617293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.481 (lit.)

bp

106 °C/4 mmHg (lit.)

density

0.898 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCCCCC1

InChI

1S/C11H20O/c12-11-9-7-5-3-1-2-4-6-8-10-11/h1-10H2

InChI key

UPOSSYJVWXLPTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cycloundecanone forms an inclusion complex with delta-cyclodextrin.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuaki Harata et al.
Chemical communications (Cambridge, England), (17)(17), 1968-1969 (2002-09-26)
The crystal structure of the delta-cyclodextrin complex with cycloundecanone shows a channel-type structure consisting of head-to-head dimer units that include four guest molecules.
H Akasaka et al.
Chemical & pharmaceutical bulletin, 48(12), 1986-1989 (2001-01-06)
The complex forming ability of delta-cyclodextrin delta-CD with 7 kinds of macrocyclic compounds (MCCs) with 8-15 carbon atoms in the ring as models of large guest molecules was studied in aqueous solution and compared with the complexation properties of alpha-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service