AABH99A47DB5
AA BLOCKS, INC.
2-phenyl-2-(thiophene-2-sulfonamido)acetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
phenyl[(2-thienylsulfonyl)amino]acetic acid
Inchi Code
1S/C12H11NO4S2/c14-12(15)11(9-5-2-1-3-6-9)13-19(16,17)10-7-4-8-18-10/h1-8,11,13H,(H,14,15)
InChI key
MQRAXDIFYJBZCA-UHFFFAOYSA-N
storage temp.
2~8 C
Purity
95%
Country of Origin
US
Product Link
https://www.aablocks.com/prod/1103529-64-1
Synonym
2-phenyl-2-(thiophene-2-sulfonamido)acetic acid
MSDS
https://www.aablocks.com/downloadSds?cas=1103529-64-1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service