Skip to Content
Merck
All Photos(1)

Documents

TP0400

Sigma-Aldrich

Total Protein Kit, Micro Pyrogallol Red Method

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for ~240 manual assays

storage temp.

2-8°C

Biochem/physiol Actions

A colorimetric assay based on pyrogallol red bound to basic amino acid groups. An increase in absorbance at 600 nm is directly proportional to the increase in protein concentration.

Kit Components Also Available Separately

Product No.
Description
SDS

  • Protein standard solution, analytical standard, 0.5 mg/mL 1 x 5

  • Total Protein Reagent, Micro Pyrogallol Red Method 2 x 120

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vishal S Vaidya et al.
American journal of physiology. Renal physiology, 290(2), F517-F529 (2005-09-22)
Sensitive and specific biomarkers are needed to detect early kidney injury. The objective of the present work was to develop a sensitive quantitative urinary test to identify renal injury in the rodent to facilitate early assessment of pathophysiological influences and
Diana V Portan et al.
BioMed research international, 2019, 7574635-7574635 (2019-04-25)
The effect of an electric field within specific intensity limits on the activity of human cells has been previously investigated. However, there are a considerable number of factors that influence the in vitro development of cell populations. In biocompatibility studies
Chandrashekar Annamalai et al.
Experimental biology and medicine (Maywood, N.J.), 245(16), 1474-1489 (2020-08-04)
This work provides in-depth insights on catalytic iron-induced cytotoxicity and the resultant triggering of endogenous vitamin D synthesis in experimental acute kidney injury. Our results reveal significantly elevated levels of catalytic iron culminating in oxidant-mediated renal injury and a concomitant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service