Merck
  • Effect of Shewanella oneidensis on the Kinetics of Fe(II)-Catalyzed Transformation of Ferrihydrite to Crystalline Iron Oxides.
Effect of Shewanella oneidensis on the Kinetics of Fe(II)-Catalyzed Transformation of Ferrihydrite to Crystalline Iron Oxides.
Environmental science & technology (2017-12-06)
Wei Xiao, Adele M Jones, Xiaomin Li, Richard N Collins, T David Waite
ZUSAMMENFASSUNG
Iron (oxyhydr)oxides are widespread in natural and engineered systems, potent adsorbents of contaminants and a source of energy for iron-reducing bacteria. Microbial reduction of iron (oxyhydr)oxides results in the formation of Fe(II) which can induce the transformation of these iron minerals, typically from less crystalline to more crystalline forms, affecting the biogeochemical cycling of iron and the behavior of any species adsorbed to the iron (oxyhydr)oxides. Factors influencing the transformation rate of the poorly crystalline iron (oxyhydr)oxide, ferrihydrite, to more crystalline forms in the presence of the iron reducing bacterium Shewanella oneidensis MR-1 are investigated under controlled laboratory conditions in this work. In particular, the amount of Fe(II) produced increased the transformation rate while increasing concentrations of the electron donor, lactate, decreased the rate. Using kinetic parameters determined from abiotic controls, the results of transformation experiments in the presence of Shewanella oneidensis were modeled with this exercise revealing that less goethite and more lepidocrocite formed than expected. Conversely, studies using the Shewanella exudate only, containing biogenic Fe(II), displayed rates of transformation that were satisfactorily modeled using these abiotic control kinetic parameters. This result suggests that the physical presence of the microbes is pivotal to the reduction in ferrihydrite transformation rate observed in the biotic experiments relative to the analogous abiotic controls.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,7-Phenanthrolin, 99%