Merck
  • Comparative analysis of serum proteome in congenital scoliosis patients with TBX6 haploinsufficiency - a first report pointing to lipid metabolism.
Comparative analysis of serum proteome in congenital scoliosis patients with TBX6 haploinsufficiency - a first report pointing to lipid metabolism.
Journal of cellular and molecular medicine (2017-09-26)
Qiankun Zhu, Nan Wu, Gang Liu, Yangzhong Zhou, Sen Liu, Jun Chen, Jiaqi Liu, Yuzhi Zuo, Zhenlei Liu, Weisheng Chen, Yixin Chen, Jia Chen, Mao Lin, Yanxue Zhao, Yang Yang, Shensgru Wang, Xu Yang, Yufen Ma, Jian Wang, Xiaoli Chen, Jianguo Zhang, Jianxiong Shen, Zhihong Wu, Guixing Qiu
ZUSAMMENFASSUNG
Congenital scoliosis (CS) is a three-dimensional deformity of the spine affecting quality of life. We have demonstrated TBX6 haploinsufficiency is the most important contributor to CS. However, the pathophysiology at the protein level remains unclear. Therefore, this study was to explore the differential proteome in serum of CS patients with TBX6 haploinsufficiency. Sera from nine CS patients with TBX6 haploinsufficiency and nine age- and gender-matched healthy controls were collected and analysed by isobaric tagged relative and absolute quantification (iTRAQ) labelling coupled with mass spectrometry (MS). In total, 277 proteins were detected and 20 proteins were designated as differentially expressed proteins, which were submitted to subsequent bioinformatics analysis. Gene Ontology classification analysis showed the biological process was primarily related to 'cellular process', molecular function 'structural molecule activity' and cellular component 'extracellular region'. IPA analysis revealed 'LXR/RXR activation' was the top pathway, which is a crucial pathway in lipid metabolism. Hierarchical clustering analysis generated two clusters. In summary, this study is the first proteomic research to delineate the total and differential serum proteins in TBX6 haploinsufficiency-caused CS. The proteins discovered in this experiment may serve as potential biomarkers for CS, and lipid metabolism might play important roles in the pathogenesis of CS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ProteoPrep® Blue Albumin & IgG Depletion Kit