Merck
  • Highly sensitive determination of diclofenac based on resin beads and a novel polyclonal antibody by using flow injection chemiluminescence competitive immunoassay.
Highly sensitive determination of diclofenac based on resin beads and a novel polyclonal antibody by using flow injection chemiluminescence competitive immunoassay.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-10-02)
Jing Shi, Mingxia Xu, Qinghui Tang, Kang Zhao, Anping Deng, Jianguo Li
ZUSAMMENFASSUNG
A novel flow injection chemiluminescence immunoassay for simple, sensitive and low-cost detection of diclofenac was established based on specific binding of antigen and antibody. Carboxylic resin beads used as solid phase carrier materials provided good biocompatibility and large surface-to-volume ratio for modifying more coating antigen. There was a competitive process between the diclofenac in solution and the immobilized coating antigen to react with the limited binding sites of the polyclonal antibody to form the immunocomplex. The second antibody labelled with horseradish peroxidase was introduced into the immunosensor and trapped by captured polyclonal antibody against diclofenac, which could effectively amplify chemiluminescence signals of luminol-PIP-H
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
5-Hydroxydiclofenac, analytical standard