Merck
Overexpression of E3 Ubiquitin Ligase Gene
Frontiers in plant science (2017-07-18)
Shuangchen Chen, Hongjiao Zhao, Mengmeng Wang, Jidi Li, Zhonghong Wang, Fenghua Wang, Airong Liu, Golam J Ahammed
ZUSAMMENFASSUNG
Ubiquitination is a common regulatory mechanism, playing a critical role in diverse cellular and developmental processes in eukaryotes. However, a few reports on the functional correlation between E3 ubiquitin ligases and reactive oxygen species (ROS) or reactive nitrogen species (RNS) metabolism in response to stress are currently available in plants. In the present study, the E3 ubiquitin ligase gene
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
S-Nitrosoglutathion, ≥97%