Merck
Polarity of varicosity initiation in central neuron mechanosensation.
The Journal of cell biology (2017-06-14)
Yuanzheng Gu, Peter Jukkola, Qian Wang, Thomas Esparza, Yi Zhao, David Brody, Chen Gu
ZUSAMMENFASSUNG
Little is known about mechanical regulation of morphological and functional polarity of central neurons. In this study, we report that mechanical stress specifically induces varicosities in the axons but not the dendrites of central neurons by activating TRPV4, a Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human TRPV4