Merck
  • Site-selective chemical modification of chymotrypsin using peptidyl derivatives bearing optically active diphenyl 1-amino-2-phenylethylphosphonate: Stereochemical effect of the diphenyl phosphonate moiety.
Site-selective chemical modification of chymotrypsin using peptidyl derivatives bearing optically active diphenyl 1-amino-2-phenylethylphosphonate: Stereochemical effect of the diphenyl phosphonate moiety.
Biopolymers (2015-12-01)
Shin Ono, Takahiko Nakai, Hirofumi Kuroda, Ryuta Miyatake, Yoshikazu Horino, Hitoshi Abe, Masahito Umezaki, Hiroshi Oyama
ZUSAMMENFASSUNG
Diphenyl (α-aminoalkyl)phosphonates act as mechanism-based inhibitors against serine proteases by forming a covalent bond with the hydroxy group of the active center Ser residue. Because the covalent bond was found to be broken and replaced by 2-pyridinaldoxime methiodide (2PAM), we employed a peptidyl derivative bearing diphenyl 1-amino-2-phenylethylphosphonate moiety (Phe(p) (OPh)2 ) to target the active site of chymotrypsin and to selectively anchor to Lys175 in the vicinity of the active site. Previously, it was reported that the configuration of the α-carbon of phosphorus in diphenyl (α-aminoalkyl)phosphonates affects the inactivation reaction of serine proteases, i.e., the (R)-enantiomeric diphenyl phosphonate is comparable to l-amino acids and it effectively reacts with serine proteases, whereas the (S)-enantiomeric form does not. In this study, we evaluated the stereochemical effect of the phosphonate moiety on the selective chemical modification. Epimeric dipeptidyl derivatives, Ala-(R or S)-Phe(p) (OPh)2 , were prepared by separation with RP-HPLC. A tripeptidyl (R)-epimer (Ala-Ala-(R)-Phe(p) (OPh)2 ) exhibited a more potent inactivation ability against chymotrypsin than the (S)-epimer. The enzyme inactivated by the (R)-epimer was more effectively reactivated with 2PAM than the enzyme inactivated by the (S)-epimer. Finally, N-succinimidyl (NHS) active ester derivatives, NHS-Suc-Ala-Ala- (R or S)-Phe(p) (OPh)2 , were prepared, and we evaluated their action when modifying Lys175 in chymotrypsin. We demonstrated that the epimeric NHS derivative that possessed the diphenyl phosphonate moiety with the (R)-configuration effectively modified Lys175 in chymotrypsin, whereas that with the (S)-configuration did not. These results demonstrate the utility of peptidyl derivatives that bear an optically active diphenyl phosphonate moiety as affinity labeling probes in protein bioconjugation. © 2015 Wiley Periodicals, Inc. Biopolymers (Pept Sci) 106: 521-530, 2016.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carbamidsäure-benzylester, 99%