Merck
Filamin A is reduced and contributes to the CASR sensitivity in human parathyroid tumors.
Journal of molecular endocrinology (2016-11-23)
Alessandra Mingione, Chiara Verdelli, Stefano Ferrero, Valentina Vaira, Vito Guarnieri, Alfredo Scillitani, Leonardo Vicentini, Gianni Balza, Edoardo Beretta, Annalisa Terranegra, Giuseppe Vezzoli, Laura Soldati, Sabrina Corbetta
ZUSAMMENFASSUNG
Parathyroid tumors display reduced sensitivity to extracellular calcium ([Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human FLNA