Merck
  • Genomic binding of PAX8-PPARG fusion protein regulates cancer-related pathways and alters the immune landscape of thyroid cancer.
Genomic binding of PAX8-PPARG fusion protein regulates cancer-related pathways and alters the immune landscape of thyroid cancer.
Oncotarget (2016-12-23)
Yanxiao Zhang, Jingcheng Yu, Vladimir Grachtchouk, Tingting Qin, Carey N Lumeng, Maureen A Sartor, Ronald J Koenig
ZUSAMMENFASSUNG
PAX8-PPARG fusion protein (PPFP) results from a t(2;3)(q13;p25) chromosomal translocation, is found in 30% of follicular thyroid carcinomas, and demonstrates oncogenic capacity in transgenic mice. A PPARG ligand, pioglitazone, is highly therapeutic in mice with PPFP thyroid cancer. However, only limited data exist to characterize the binding sites and oncogenic function of PPFP, or to explain the observed therapeutic effect of pioglitazone. Here we used our previously characterized transgenic mouse model of PPFP follicular thyroid carcinoma to identify PPFP binding sites in vivo using ChIP-seq, and to distinguish genes and pathways regulated directly or indirectly by PPFP with and without pioglitazone treatment via integration with RNA-seq data. PPFP bound to DNA regions containing the PAX8 and/or the PPARG motif, near genes involved in lipid metabolism, the cell cycle, apoptosis, and cell motility; the binding site distribution was highly concordant with our previous study in a rat PCCL3 cell line. Most strikingly, pioglitazone induced an immune cell infiltration including macrophages and T cells only in the presence of PPFP, which may be central to its therapeutic effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
c-Myc Peptide, lyophilized powder