Merck
A Fluoro-Chromogenic Sensor Based on Organic Molecular Framework for Cu
Journal of fluorescence (2016-10-13)
Veikko Uahengo, Yan Zhang, Bi Xiong, Pingping Zhao, Ping Cai, Lydia Rhyman, Ponnadurai Ramasami, Kai Hu, Gongzhen Cheng
ZUSAMMENFASSUNG
The 2,2'-dinaphtholazobenzene molecular framework (P) was designed, synthesized and characterized. Its absorption and fluorescence properties revealed that P is a dual sensor for copper ions (Cu
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Hydroxy-2-naphthaldehyde, AldrichCPR