Merck
  • Novel hydrolytic de-methylthiolation of the s-triazine herbicide prometryn by Leucobacter sp. JW-1.
Novel hydrolytic de-methylthiolation of the s-triazine herbicide prometryn by Leucobacter sp. JW-1.
The Science of the total environment (2016-11-22)
Junwei Liu, Rimao Hua, Pei Lv, Jun Tang, Yi Wang, Haiqun Cao, Xiangwei Wu, Qing X Li
ZUSAMMENFASSUNG
s-Triazine herbicides have been widely used in recent decades and caused serious concern over contamination of groundwater, surface water and soil. A novel bacterial strain JW-1 was isolated from activated sludge and identified as Leucobacter sp. based on comparative morphology, physiological characteristics and comparison of the 16S rDNA gene sequence. JW-1 was capable of using methylthio-s-triazine prometryn as a sole source of carbon and energy in pure culture. Favorable conditions for prometryn degradation were found at pH7.0-9.0 and temperature of 37-42°C. The degradation half-life of prometryn at 50mgL
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Terbumeton, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.