Merck
  • Ex vivo desequestration of Plasmodium falciparum-infected erythrocytes from human placenta by chondroitin sulfate A.
Ex vivo desequestration of Plasmodium falciparum-infected erythrocytes from human placenta by chondroitin sulfate A.
Infection and immunity (1999-11-24)
J Gysin, B Pouvelle, N Fievet, A Scherf, C Lépolard
ZUSAMMENFASSUNG
We performed ex vivo experiments with Plasmodium falciparum-infected human placentas from primi- and multigravida women from Cameroon. All women, independent of their gravida status, had anti-chondroitin sulfate A (CSA) adhesion antibodies which cross-reacted with heterologous strains, such as FCR3 and Palo-Alto(FUP)1, which were selected for CSA binding. These antibodies, directed against the surface of infected erythrocytes obtained by flushing with CSA (IRBC(CSA)), were restricted to the immunoglobulin G3 isotypes. Massive desequestration of parasites was achieved with soluble CSA but not with anti-ICAM-1 and anti-CD36 monoclonal antibodies. All of the CSA-flushed parasites were analyzed immediately by using in vitro assays of binding to Saimiri brain endothelial cells (SBEC) expressing various adhesion receptors. Parasites derived from all six placentas displayed the CSA adhesion phenotype. However, only partial inhibition of adhesion was observed in the presence of soluble CSA or when Sc1D SBEC were treated with chondroitinase ABC. These results suggest that an additional adhesive molecule of IRBC(CSA) which binds to an unidentified receptor is present in the placenta. This new phenotype was lost once the parasites adapted to in vitro culture. We observed additional differences in the CSA adhesion phenotype between placental parasites and in vitro-cultured parasites panned on endothelial cells carrying CSA. The minimum size of fractionated CSA required for a significant inhibition of placental IRBC(CSA) adhesion to Sc1D cells was 1 to 2 kDa, which contrasts with the 4-kDa size necessary to reach equivalent levels of inhibition with panned IRBC(CSA) of this phenotype. All placental IRBC(CSA) cytoadhered to Sc17 SBEC, which express only the CSA receptor. Panning of IRBC(CSA) on these cells resulted in a significant quantitative increase of IRBC cytoadhering to the CSA of Sc1D cells but did not change their capacity for adhesion to CSA on normal placenta cryosections. Our results indicate that the CSA binding phenotype is heterogeneous and that several distinct genes may encode P. falciparum-CSA ligands with distinct binding properties.