Merck
Proteomic based approach for characterizing 4-hydroxy-2-nonenal induced oxidation of buffalo (
Proteome science (2016-11-29)
Naveena B Maheswarappa, K Usha Rani, Y Praveen Kumar, Vinayak V Kulkarni, Srikanth Rapole
ZUSAMMENFASSUNG
Myoglobin (Mb) is a sarcoplasmic heme protein primarily responsible for meat color and its chemistry is species specific. 4-hydroxy-2-nonenal (HNE) is a cytotoxic lipid derived aldehyde detected in meat and was reported to covalently adduct with nucleophilic histidine residues of Mb and predispose it to greater oxidation. However, no literature is available on characterization of lipid oxidation induced oxidation of Indian water buffalo ( Present study characterize the Mb extracted from water buffalo and goat cardiac muscles using two-dimensional gel electrophoresis (2DE), OFFGEL electrophoresis and mass spectrometry (MS). Purified buffalo and goat bright red oxymyoglobin were reacted with HNE in-vitro at physiological pH (7.4) and temperature (37 °C) conditions and the formation of oxidised brown metmyoglobin was measured. The Mb-HNE adducts were detected using MALDI-TOF MS, whereas specific sites of adduction was determined using ESI-QTOF MS/MS. Purified buffalo and goat Mb samples revealed a molecular mass of 17,043.6 and 16,899.9 Daltons, respectively. The 2DE analysis exhibited 65 (sarcoplasmic protein extract) and 6 (pure Mb) differentially expressed ( The study demonstrated better redox stability of buffalo Mb than goat Mb. Our findings confirm the hypothesis that relative effect of HNE was greater for Mb's with 12 ± 1 HIS residues than Mb's with 9 HIS residues and helps meat processors in developing species-specific processing strategies to reduce the color variability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Allzweck-Viskositätsstandard, durch UKAS ISO/IEC17025 und ISO 17034 zertifiziert, viscosity 392 mPa.s (25 °C)