Merck
Doxorubicin and resveratrol co-delivery nanoparticle to overcome doxorubicin resistance.
Scientific reports (2016-10-13)
Yuan Zhao, Meng-Lei Huan, Miao Liu, Ying Cheng, Yang Sun, Han Cui, Dao-Zhou Liu, Qi-Bing Mei, Si-Yuan Zhou
ZUSAMMENFASSUNG
With the extensive application of doxorubicin (DOX), DOX resistance has become one of the main obstacles to the effective treatment of breast cancer. In this paper, DOX and resveratrol (RES) were co-encapsulated in a modified PLGA nanoparticle (NPS) to overcome the DOX resistance. CLSM results indicated that DOX and RES were simultaneously delivered into the nucleus of DOX-resistant human breast cancer cells by DOX/RES-loaded NPS. Consequently, DOX/RES-loaded NPS showed significant cytotoxicity on MDA-MB-231/ADR cells and MCF-7/ADR cells. Furthermore, DOX/RES-loaded NPS could overcome DOX resistance by inhibiting the expression of drug resistance-related protein such as P-gp, MRP-1 and BCRP, and induce apoptosis through down-regulating the expression of NF-κB and BCL-2. In tumor-bearing mice, DOX/RES-loaded NPS mainly delivered DOX and RES to tumor tissue. Compared with free DOX, DOX/RES-loaded NPS significantly inhibited the DOX-resistant tumor growth in tumor-bearing mice without causing significant systemic toxicity. In a word, DOX/RES-loaded NPS could overcome the DOX resistance and had the potential in the treatment of DOX-resistant breast cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Methoxy-benzoylchlorid, 99%