Merck
Breast cancer cell line proliferation blocked by the Src-related Rak tyrosine kinase.
International journal of cancer (2003-02-06)
Tanya Meyer, LiHui Xu, Jinli Chang, Edison T Liu, Rolf J Craven, William G Cance
ZUSAMMENFASSUNG
Rak is a 54 kDa protein tyrosine kinase originally isolated from breast cancer cells and expressed in epithelial cells. It resembles the protooncogene Src structurally but lacks an amino-terminal myristylation site and localizes to the nuclear and perinuclear regions of the cell. We report here that expression of Rak in 2 different breast cancer cell lines inhibits growth and causes G(1) arrest of the cell cycle. This growth inhibition is kinase-dependent but does not require the Rak SH2 or SH3 domain. Rak also binds to the pRb tumor-suppressor protein but inhibits growth even in cells that lack pRb. These results suggest that Rak regulates cell growth by phosphorylating perinuclear proteins and has a function that is distinct from the Src-related kinase family.