- An efficient two-step total synthesis of the quaterpyridine nemertelline.
An efficient two-step total synthesis of the quaterpyridine nemertelline.
The Journal of organic chemistry (2003-12-20)
Alexandre Bouillon, Anne Sophie Voisin, Audrey Robic, Jean-Charles Lancelot, Valérie Collot, Sylvain Rault
PMID14682721
ZUSAMMENFASSUNG
Regioselective and univocal Suzuki cross-coupling reactions performed on halopyridinyl boronic acids provide a flexible and versatile route to a multigram scale synthesis of 2,2'-dichloro-3,4'-bipyridine 14, which allows couplings with excess pyridin-3-yl boronic acid to give a new and efficient two-step rapid synthesis of nemertelline, the quaterpyridine neurotoxin isolated from a Hoplonemertine sea worm.