Merck
  • Synthesis, structure, and chemistry of new, mixed group 14 and 16 heterocycles: nucleophile-induced ring contraction of mesocyclic dications.
Synthesis, structure, and chemistry of new, mixed group 14 and 16 heterocycles: nucleophile-induced ring contraction of mesocyclic dications.
Journal of the American Chemical Society (2006-11-16)
Eric Block, Evgeny V Dikarev, Richard S Glass, Jin Jin, Bo Li, Xiaojie Li, Shao-Zhong Zhang
ZUSAMMENFASSUNG
More than 40 new 4- to 12-membered ring heterocycles containing various combinations of Group 14 and 16 elements Si, Sn, S, Se, and Te have been synthesized and fully characterized. Synthesis of these small-ring as well as medium-ring (mesocyclic) heterocycles from alpha,omega-dihalides is facilitated by the presence of gem-dialkylsilyl and gem-dialkylstannyl groups in the precursors. Conformations of several of the new ring systems in the solid state have been determined by X-ray crystal structure analysis. Oxidation of mixed S(Se, Te)/Si eight-membered ring mesocycles with NOPF6 or Br2 gives dications or a bicyclic dibromide, respectively, which can be characterized by NMR methods. On treatment with nucleophiles, mesocyclic dications, or the corresponding radical cations undergo ring contraction, giving five- or six-membered ring heterocycles. Photolysis of a S/Se four-membered ring heterocycle gives selenoformaldehyde, trapped in 80% yield with 2,3-dimethyl-1,3-butadiene.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumselencyanat, reagent grade, 97%