Merck
  • Disposition of pravastatin sodium, a tissue-selective HMG-CoA reductase inhibitor, in healthy subjects.
Disposition of pravastatin sodium, a tissue-selective HMG-CoA reductase inhibitor, in healthy subjects.
British journal of clinical pharmacology (1990-02-01)
S M Singhvi, H Y Pan, R A Morrison, D A Willard
ZUSAMMENFASSUNG
Pravastatin sodium, a competitive inhibitor of HMG-CoA reductase, is a new orally effective hypocholesterolaemic agent. In a two-way crossover study, eight healthy male subjects each received an intravenous and an oral dose of [14C]-pravastatin sodium. The oral absorption of [14C] activity from pravastatin sodium was about 34% and the oral bioavailability was about 18%, suggesting first-pass metabolism of pravastatin. After the intravenous dose, the recovery of radioactivity averaged 60% and 34% in urine and faeces, respectively. Corresponding values were 20% (urine) and 71% (faeces) for the oral dose. The estimated average plasma elimination half-life of pravastatin was 0.8 and 1.8 h for the intravenous and oral routes, respectively. The average values for total and renal clearances were 13.5 and 6.3 ml min-1 kg-1, respectively, and the steady-state volume of distribution averaged 0.51 kg-1. These results suggest that both kidney and liver are important sites of elimination for pravastatin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pravastatin sodium salt hydrate, ≥98% (HPLC), powder