Merck
  • En route to osmium analogues of KP1019: synthesis, structure, spectroscopic properties and antiproliferative activity of trans-[Os(IV)Cl4(Hazole)2].
En route to osmium analogues of KP1019: synthesis, structure, spectroscopic properties and antiproliferative activity of trans-[Os(IV)Cl4(Hazole)2].
Inorganic chemistry (2011-07-12)
Gabriel E Büchel, Iryna N Stepanenko, Michaela Hejl, Michael A Jakupec, Bernhard K Keppler, Vladimir B Arion
ZUSAMMENFASSUNG
By controlled Anderson type rearrangement reactions complexes of the general formula trans-[Os(IV)Cl(4)(Hazole)(2)], where Hazole = 1H-pyrazole, 2H-indazole, 1H-imidazole, and 1H-benzimidazole, have been synthesized. Note that 2H-indazole tautomer stabilization in trans-[Os(IV)Cl(4)(2H-indazole)(2)] is unprecedented in coordination chemistry of indazole. The metal ion in these compounds possesses the same coordination environment as ruthenium(III) in (H(2)ind)[Ru(III)Cl(4)(Hind)(2)], where Hind = 1H-indazole, (KP1019), an investigational anticancer drug in phase I clinical trials. These osmium(IV) complexes are appropriate precursors for the synthesis of osmium(III) analogues of KP1019. In addition the formation of an adduct of trans-[Os(IV)Cl(4)(Hpz)(2)] with cucurbit[7]uril is described. The compounds have been comprehensively characterized by elemental analysis, EI and ESI mass spectrometry, spectroscopy (IR, UV-vis, 1D and 2D NMR), cyclic voltammetry, and X-ray crystallography. Their antiproliferative acitivity in the human cancer cell lines CH1 (ovarian carcinoma), A549 (nonsmall cell lung carcinoma), and SW480 (colon carcinoma) is reported.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indazol, 98%